InfinityLab Poroshell 120 EC-C8, 3, 250 mm, 4 um, solvent saver LC column, with column ID. Specifications:- Carbon Load: 5 %. Endcap ped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 3 mm. LC Column Brand: InfinityLab Poroshell 1 * 1 Pce
Fournisseur: Agilent
InfinityLab Poroshell 120 EC-C8, 3, 250 mm, 4 um, solvent saver LC column, with column ID. Specifications:- Carbon Load: 5 %. Endcap ped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 3 mm. LC Column Brand: InfinityLab Poroshell 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
InfinityLab Poroshell 120 EC-C8, 3, 250 mm, 4 um, solvent saver LC column, with column ID. Specifications:- Carbon Load: 5 %. Endcap ped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 3 mm. LC Column Brand: InfinityLab Poroshell 1 * 1 Pce HEWL690970-306T 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou