InfinityLab Poroshell 120 Bonus-RP, 3.0 x 150 mm, 2.7 um, solvent saver LC column, with column ID. Specifications:- Carbon Load: 7.5 %. Endcapped: Triple. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 3 mm. LC Column Brand: Infinity 1 * 1 Pce
Fournisseur: Agilent
InfinityLab Poroshell 120 Bonus-RP, 3.0 x 150 mm, 2.7 um, solvent saver LC column, with column ID. Specifications:- Carbon Load: 7.5 %. Endcapped: Triple. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 3 mm. LC Column Brand: Infinity 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
InfinityLab Poroshell 120 Bonus-RP, 3.0 x 150 mm, 2.7 um, solvent saver LC column, with column ID. Specifications:- Carbon Load: 7.5 %. Endcapped: Triple. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 3 mm. LC Column Brand: Infinity 1 * 1 Pce HEWL693968-301T 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou