InfinityLab Poroshell HPH-C18, 3.0 x 150 mm, 2.7 um, solvent saver LC column, with column ID. Specifications:- Endcapped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 3 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column 1 * 1 Pce
Fournisseur: Agilent
InfinityLab Poroshell HPH-C18, 3.0 x 150 mm, 2.7 um, solvent saver LC column, with column ID. Specifications:- Endcapped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 3 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
InfinityLab Poroshell HPH-C18, 3.0 x 150 mm, 2.7 um, solvent saver LC column, with column ID. Specifications:- Endcapped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 3 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column 1 * 1 Pce HEWL693975-502T 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou