Specifications:- Carbon Load: 10 %. Endcapped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column Maximum Temperature: 60 C . LC Column Phase: EC-C18. LC Platform: Low Di 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Carbon Load: 10 %. Endcapped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column Maximum Temperature: 60 C . LC Column Phase: EC-C18. LC Platform: Low Di 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Carbon Load: 10 %. Endcapped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column Maximum Temperature: 60 C . LC Column Phase: EC-C18. LC Platform: Low Di 1 * 1 Pce HEWL695775-302 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou