InfinityLab Poroshell HPH-C8, 2.1 x 100 mm, 2.7 um, narrow bore LC column, with column ID. Specifications:- Endcapped: Yes. Guard Co lumn: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column 1 * 1 Pce
Fournisseur: Agilent
InfinityLab Poroshell HPH-C8, 2.1 x 100 mm, 2.7 um, narrow bore LC column, with column ID. Specifications:- Endcapped: Yes. Guard Co lumn: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
InfinityLab Poroshell HPH-C8, 2.1 x 100 mm, 2.7 um, narrow bore LC column, with column ID. Specifications:- Endcapped: Yes. Guard Co lumn: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column 1 * 1 Pce HEWL695775-706T 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou