InfinityLab Poroshell 120 HILIC, 4.6 x 100 mm, 4 um, analytical LC column, with column ID. Specifications:- Carbon Load: 0 %. Endcap ped: No. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 4.6 mm. LC Column Brand: InfinityLab Poroshel 1 * 1 Pce
Fournisseur: Agilent
InfinityLab Poroshell 120 HILIC, 4.6 x 100 mm, 4 um, analytical LC column, with column ID. Specifications:- Carbon Load: 0 %. Endcap ped: No. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 4.6 mm. LC Column Brand: InfinityLab Poroshel 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
InfinityLab Poroshell 120 HILIC, 4.6 x 100 mm, 4 um, analytical LC column, with column ID. Specifications:- Carbon Load: 0 %. Endcap ped: No. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 4.6 mm. LC Column Brand: InfinityLab Poroshel 1 * 1 Pce HEWL695970-901T 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou