InfinityLab Poroshell HPH-C8, 2.1 , 50 mm, 2.7 um, narrow bore LC column, with column ID. Specifications:- Endcapped: Yes. Guard Col umn: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column M 1 * 1 Pce
Fournisseur: Agilent
InfinityLab Poroshell HPH-C8, 2.1 , 50 mm, 2.7 um, narrow bore LC column, with column ID. Specifications:- Endcapped: Yes. Guard Col umn: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column M 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
InfinityLab Poroshell HPH-C8, 2.1 , 50 mm, 2.7 um, narrow bore LC column, with column ID. Specifications:- Endcapped: Yes. Guard Col umn: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column M 1 * 1 Pce HEWL699775-706T 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou