InfinityLab Poroshell 120 EC-C8, 2.1 x 50 mm, 2.7 um, narrow bore LC column, with column ID. Specifications:- Carbon Load: 5 %. Endc apped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poros 1 * 1 Pce
Fournisseur: Agilent
InfinityLab Poroshell 120 EC-C8, 2.1 x 50 mm, 2.7 um, narrow bore LC column, with column ID. Specifications:- Carbon Load: 5 %. Endc apped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poros 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
InfinityLab Poroshell 120 EC-C8, 2.1 x 50 mm, 2.7 um, narrow bore LC column, with column ID. Specifications:- Carbon Load: 5 %. Endc apped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poros 1 * 1 Pce HEWL699775-906T 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou