InfinityLab Poroshell 120 Phenyl-Hexyl, 2.1, 50 mm, 2.7 um, narrow bore LC column, with column ID. Specifications:- Carbon Load: 8 % . Endcapped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab 1 * 1 Pce
Fournisseur: Agilent
InfinityLab Poroshell 120 Phenyl-Hexyl, 2.1, 50 mm, 2.7 um, narrow bore LC column, with column ID. Specifications:- Carbon Load: 8 % . Endcapped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
InfinityLab Poroshell 120 Phenyl-Hexyl, 2.1, 50 mm, 2.7 um, narrow bore LC column, with column ID. Specifications:- Carbon Load: 8 % . Endcapped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab 1 * 1 Pce HEWL699775-912T 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou