InfinityLab Poroshell HPH-C8, 4.6 x 50 mm, 4 um, analytical LC column, with column ID. Specifications:- Endcapped: Yes. Guard Column : No. Hardware: Conventional. Inner Diameter (ID): 4.6 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column Maxi 1 * 1 Pce
Fournisseur: Agilent
InfinityLab Poroshell HPH-C8, 4.6 x 50 mm, 4 um, analytical LC column, with column ID. Specifications:- Endcapped: Yes. Guard Column : No. Hardware: Conventional. Inner Diameter (ID): 4.6 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column Maxi 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
InfinityLab Poroshell HPH-C8, 4.6 x 50 mm, 4 um, analytical LC column, with column ID. Specifications:- Endcapped: Yes. Guard Column : No. Hardware: Conventional. Inner Diameter (ID): 4.6 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column Maxi 1 * 1 Pce HEWL699970-706T 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou