InfinityLab Poroshell HPH-C8, 3.0 x 50 mm, 2.7 um, solvent saver LC column, with column ID. Specifications:- Endcapped: Yes. Guard C olumn: No. Hardware: Conventional. Inner Diameter (ID): 3 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column M 1 * 1 Pce
Fournisseur: Agilent
InfinityLab Poroshell HPH-C8, 3.0 x 50 mm, 2.7 um, solvent saver LC column, with column ID. Specifications:- Endcapped: Yes. Guard C olumn: No. Hardware: Conventional. Inner Diameter (ID): 3 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column M 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
InfinityLab Poroshell HPH-C8, 3.0 x 50 mm, 2.7 um, solvent saver LC column, with column ID. Specifications:- Endcapped: Yes. Guard C olumn: No. Hardware: Conventional. Inner Diameter (ID): 3 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column M 1 * 1 Pce HEWL699975-506T 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou