ICP-OES nebulizers and supplies for PerkinElmer
Fournisseur: Agilent
8003-0320 8003-0322 8003-0324 8003-0326 8003-0327 8003-0328 8003-0335 8003-0454 8003-0461 8003-0464 8003-0951 8003-0964 8003-0974
HEWL8003-0320EA 849 EUR InStock
HEWL8003-0320 HEWL8003-0322 HEWL8003-0324 HEWL8003-0326 HEWL8003-0327 HEWL8003-0328 HEWL8003-0335 HEWL8003-0454 HEWL8003-0461 HEWL8003-0464 HEWL8003-0951 HEWL8003-0964 HEWL8003-0974
ICP-OES nebulizers and supplies for PerkinElmer
Matériel de chromatographie Chromatographie, Pièces de rechange
These supplies can enhance the performance and productivity of PerkinElmer spectroscopy systems.
  • Manufactured to fine tolerances for reproducible and stable sample uptake
  • Tight specifications deliver fine droplets for best signal intensity and optimum signal to noise
  • Fewer repeats means less downtime and improved productivity
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the stand behind warranty that guarantees the parts will not cause instrument failure or downtime
  • Parts qualified on a genuine PerkinElmer system
  • Many parts in packaging designed to be more durable, sturdy and provide the best protection during shipping to ensure the part arrives with no damage
Nebulizers for PerkinElmer ICP-OES systems have tight specifications for delivering fine droplets for the best signal intensity and optimum signal to noise. Choose from GemTip, GemCone, Meinhard, or inert MiraMist. The unique OneNeb series 2 nebulizer improves performance compared to glass concentric and inert types used with the PerkinElmer Optima ICP-OES instruments. The OneNeb series 2 is a direct replacement for most other nebulizers and is up to four times more sensitive. The OneNeb is also rugged and durable and provides superb tolerance to high levels of dissolved solids and aggressive acids.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité