Spray chambers and supplies for PerkinElmer
Fournisseur: Agilent
8003-0329 8003-0330 8003-0331 8003-0332 8003-0333 8003-0334 8003-0457 8003-0460 8003-0462 8003-0463 8003-0817 8003-0953 8003-0963
HEWL8003-0329EA 947 EUR InStock
HEWL8003-0329 HEWL8003-0330 HEWL8003-0331 HEWL8003-0332 HEWL8003-0333 HEWL8003-0334 HEWL8003-0457 HEWL8003-0460 HEWL8003-0462 HEWL8003-0463 HEWL8003-0817 HEWL8003-0953 HEWL8003-0963
Spray chambers and supplies for PerkinElmer
Matériel de chromatographie Chromatographie, Pièces de rechange
These spray chambers and supplies are fully compatible with your PerkinElmer ICP-OES.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Parts qualified on a genuine PerkinElmer system
  • Many parts in Agilent packaging designed to be more durable, sturdy and provide the best protection during shipping to ensure the part arrives with no damage
The single pass glass cyclonic spray chamber for good signal to noise performance with excellent washout. Alternatively, the double pass glass cyclonic spray chamber provides improved plasma stability with organic and high TDS samples.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité