Spray chambers for PerkinElmer
Fournisseur: Agilent
8003-0496 8003-0501 8003-0514 8003-0515 8003-0516 8003-0517 8003-0518 8003-0519 8003-0520 8003-0521 8003-0522 8003-0523 8003-0524 8003-0588 8003-0605 8003-0614 8003-0946 8003-0947 8003-0948
HEWL8003-0496EA 140 EUR InStock
HEWL8003-0496 HEWL8003-0501 HEWL8003-0514 HEWL8003-0515 HEWL8003-0516 HEWL8003-0517 HEWL8003-0518 HEWL8003-0519 HEWL8003-0520 HEWL8003-0521 HEWL8003-0522 HEWL8003-0523 HEWL8003-0524 HEWL8003-0588 HEWL8003-0605 HEWL8003-0614 HEWL8003-0946 HEWL8003-0947 HEWL8003-0948
Spray chambers for PerkinElmer
Matériel de chromatographie Chromatographie, Pièces de rechange
These spray chambers and supplies are fully compatible with your PerkinElmer ICP-MS.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Parts qualified on a genuine PerkinElmer system
  • Many parts in Agilent packaging designed to be more durable, sturdy and provide the best protection during shipping to ensure the part arrives with no damage
The single pass glass cyclonic spray chamber for good signal to noise performance with excellent washout. Alternatively, the double pass glass cyclonic spray chamber provides improved plasma stability with organic and high TDS samples.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité