Torch injectors for PerkinElmer
Fournisseur: Agilent
8003-0530 8003-0531 8003-0532 8003-0533 8003-0534 8003-0535 8003-0538 8003-0539 8003-0540 8003-0541 8003-0542 8003-0543 8003-0544 8003-0590 8003-0594
HEWL8003-0530EA 265 EUR InStock
HEWL8003-0530 HEWL8003-0531 HEWL8003-0532 HEWL8003-0533 HEWL8003-0534 HEWL8003-0535 HEWL8003-0538 HEWL8003-0539 HEWL8003-0540 HEWL8003-0541 HEWL8003-0542 HEWL8003-0543 HEWL8003-0544 HEWL8003-0590 HEWL8003-0594
Torch injectors for PerkinElmer
Matériel de chromatographie Chromatographie, Pièces de rechange
These torch injectors for PerkinElmer ICP-MS are available in a wide range of types.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Parts qualified on a genuine PerkinElmer system
  • Many parts in Agilent packaging designed to be more durable, sturdy and provide the best protection during shipping to ensure the part arrives with no damage
Quartz injectors are used for volatile organic solvent introduction and recommended for a wide range of non-HF applications. Sapphire injectors are used for environmental, geochemistry, and HF applications.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité