Specifications:- Manufacturer Compatibility: CTC. Needle Length: 57 mm. Needle Point Style: Cone. Needle Type: Fixed. Syringe Needle Gauge: 26. Technique: GC & GC/MS. Volume: 0.5 uL. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Manufacturer Compatibility: CTC. Needle Length: 57 mm. Needle Point Style: Cone. Needle Type: Fixed. Syringe Needle Gauge: 26. Technique: GC & GC/MS. Volume: 0.5 uL. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Manufacturer Compatibility: CTC. Needle Length: 57 mm. Needle Point Style: Cone. Needle Type: Fixed. Syringe Needle Gauge: 26. Technique: GC & GC/MS. Volume: 0.5 uL. 1 * 1 Pce HEWL8010-1300 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou