Specifications:- Needle Inside Diameter (ID): 0.686 mm. Needle Length: 85 mm. Needle Material: Stainless steel. Needle Outside Diame ter (OD): 1.067 mm. Needle Point Style: Side Hole. Needle Type: Removable. Syringe Needle Gauge: 19. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Needle Inside Diameter (ID): 0.686 mm. Needle Length: 85 mm. Needle Material: Stainless steel. Needle Outside Diame ter (OD): 1.067 mm. Needle Point Style: Side Hole. Needle Type: Removable. Syringe Needle Gauge: 19. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Needle Inside Diameter (ID): 0.686 mm. Needle Length: 85 mm. Needle Material: Stainless steel. Needle Outside Diame ter (OD): 1.067 mm. Needle Point Style: Side Hole. Needle Type: Removable. Syringe Needle Gauge: 19. 1 * 1 Pce HEWL8010-1399 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou