ZORBAX RRHD Bonus-RP, 80A, 2.1 mm x 5 mm, 1.8 um, 1200 bar pressure limit, UHPLC guard, 3/pk. Specifications:- Carbon Load: 9.5 %. E ndcapped: Triple. Guard Column: Yes. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: ZORBAX. LC 1 * 3 Pce
Fournisseur: Agilent
ZORBAX RRHD Bonus-RP, 80A, 2.1 mm x 5 mm, 1.8 um, 1200 bar pressure limit, UHPLC guard, 3/pk. Specifications:- Carbon Load: 9.5 %. E ndcapped: Triple. Guard Column: Yes. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: ZORBAX. LC 1 * 3 Pce
COMMANDE
Développer tout / Réduire tout
ZORBAX RRHD Bonus-RP, 80A, 2.1 mm x 5 mm, 1.8 um, 1200 bar pressure limit, UHPLC guard, 3/pk. Specifications:- Carbon Load: 9.5 %. E ndcapped: Triple. Guard Column: Yes. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: ZORBAX. LC 1 * 3 Pce HEWL821725-937 1 * 3 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou