Specifications:- Carbon Load: 4 %. Endcapped: No. Guard Column: Yes. Hardware: Conventional. Inner Diameter (ID): 3 mm. LC Column Br and: ZORBAX. LC Column Maximum Temperature: 80 C at pH 1-6 40 C at pH 6-8 . LC Column Phase: SB-CN. LC Platform: UH 1 * 3 Pce
Fournisseur: Agilent
Specifications:- Carbon Load: 4 %. Endcapped: No. Guard Column: Yes. Hardware: Conventional. Inner Diameter (ID): 3 mm. LC Column Br and: ZORBAX. LC Column Maximum Temperature: 80 C at pH 1-6 40 C at pH 6-8 . LC Column Phase: SB-CN. LC Platform: UH 1 * 3 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Carbon Load: 4 %. Endcapped: No. Guard Column: Yes. Hardware: Conventional. Inner Diameter (ID): 3 mm. LC Column Br and: ZORBAX. LC Column Maximum Temperature: 80 C at pH 1-6 40 C at pH 6-8 . LC Column Phase: SB-CN. LC Platform: UH 1 * 3 Pce HEWL823750-934 1 * 3 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou