Specifications:- Endcapped: No. Guard Column: Yes. Hardware: Conventional. Inner Diameter (ID): 3 mm. LC Column Brand: ZORBAX. LC Co lumn Maximum Temperature: 80 C at pH 1-6 40 C at pH 6-8 . LC Column Phase: SB-Aq. LC Platform: UHPLC Low Dispersion 1 * 3 Pce
Fournisseur: Agilent
Specifications:- Endcapped: No. Guard Column: Yes. Hardware: Conventional. Inner Diameter (ID): 3 mm. LC Column Brand: ZORBAX. LC Co lumn Maximum Temperature: 80 C at pH 1-6 40 C at pH 6-8 . LC Column Phase: SB-Aq. LC Platform: UHPLC Low Dispersion 1 * 3 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Endcapped: No. Guard Column: Yes. Hardware: Conventional. Inner Diameter (ID): 3 mm. LC Column Brand: ZORBAX. LC Co lumn Maximum Temperature: 80 C at pH 1-6 40 C at pH 6-8 . LC Column Phase: SB-Aq. LC Platform: UHPLC Low Dispersion 1 * 3 Pce HEWL823750-936 1 * 3 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou