Specifications:- Hardware: SS. Inner Diameter (ID): 2.1 mm. LC Column Brand: ZORBAX RRHD 300SB-Diphenyl. LC Column Maximum Temperatu re: 80 C . LC Column Phase: 300-Diphenyl. LC Platform: Analytical - Stainless Steel, UHPLC ( 600 bar). Length: 150 mm 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Hardware: SS. Inner Diameter (ID): 2.1 mm. LC Column Brand: ZORBAX RRHD 300SB-Diphenyl. LC Column Maximum Temperatu re: 80 C . LC Column Phase: 300-Diphenyl. LC Platform: Analytical - Stainless Steel, UHPLC ( 600 bar). Length: 150 mm 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Hardware: SS. Inner Diameter (ID): 2.1 mm. LC Column Brand: ZORBAX RRHD 300SB-Diphenyl. LC Column Maximum Temperatu re: 80 C . LC Column Phase: 300-Diphenyl. LC Platform: Analytical - Stainless Steel, UHPLC ( 600 bar). Length: 150 mm 1 * 1 Pce HEWL863750-944 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou