Specifications:- Capillary Column Phase: CP-Volamine. Capillary Tubing: Fused Silica. GC Column Dim: 60 m x . GC Column Format: Intu vo. Inner Diameter (ID): 0.32 mm. Length: 60 cm. Length (m): 60 m. Polarity: Low Polarity. With Smart Key: Yes. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Capillary Column Phase: CP-Volamine. Capillary Tubing: Fused Silica. GC Column Dim: 60 m x . GC Column Format: Intu vo. Inner Diameter (ID): 0.32 mm. Length: 60 cm. Length (m): 60 m. Polarity: Low Polarity. With Smart Key: Yes. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Capillary Column Phase: CP-Volamine. Capillary Tubing: Fused Silica. GC Column Dim: 60 m x . GC Column Format: Intu vo. Inner Diameter (ID): 0.32 mm. Length: 60 cm. Length (m): 60 m. Polarity: Low Polarity. With Smart Key: Yes. 1 * 1 Pce HEWLCP7448-INT 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou