Specifications:- Capillary Column Phase: Select PAH. Capillary Tubing: Fused Silica. Film Thickness: 0.15 um. GC Column Dim: 30 m x 0.15 um. GC Column Format: Intuvo. Inner Diameter (ID): 0.25 mm. Length: 30 mm. Length (m): 30 m. Polarity: Low Polari 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Capillary Column Phase: Select PAH. Capillary Tubing: Fused Silica. Film Thickness: 0.15 um. GC Column Dim: 30 m x 0.15 um. GC Column Format: Intuvo. Inner Diameter (ID): 0.25 mm. Length: 30 mm. Length (m): 30 m. Polarity: Low Polari 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Capillary Column Phase: Select PAH. Capillary Tubing: Fused Silica. Film Thickness: 0.15 um. GC Column Dim: 30 m x 0.15 um. GC Column Format: Intuvo. Inner Diameter (ID): 0.25 mm. Length: 30 mm. Length (m): 30 m. Polarity: Low Polari 1 * 1 Pce HEWLCP7462-INT 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou