Specifications:- Capillary Column Phase: CP-TAP CB. Capillary Tubing: Fused Silica. Film Thickness: 0.1 um. GC Column Dim: 25 m x 0. 1 um. GC Column Format: Intuvo. Inner Diameter (ID): 0.25 mm. Length: 25 m. Length (m): 25 m. With Smart Key: Yes. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Capillary Column Phase: CP-TAP CB. Capillary Tubing: Fused Silica. Film Thickness: 0.1 um. GC Column Dim: 25 m x 0. 1 um. GC Column Format: Intuvo. Inner Diameter (ID): 0.25 mm. Length: 25 m. Length (m): 25 m. With Smart Key: Yes. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Capillary Column Phase: CP-TAP CB. Capillary Tubing: Fused Silica. Film Thickness: 0.1 um. GC Column Dim: 25 m x 0. 1 um. GC Column Format: Intuvo. Inner Diameter (ID): 0.25 mm. Length: 25 m. Length (m): 25 m. With Smart Key: Yes. 1 * 1 Pce HEWLCP7483-INT 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou