Specifications:- Capillary Column Phase: CP-Sil 88. Capillary Tubing: Fused Silica. Film Thickness: 0.2 um. GC Column Dim: 60 m x 0. 2 um. GC Column Format: Intuvo. Inner Diameter (ID): 0.25 mm. Length: 60 m. Length (m): 60 m. Polarity: High Polarity 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Capillary Column Phase: CP-Sil 88. Capillary Tubing: Fused Silica. Film Thickness: 0.2 um. GC Column Dim: 60 m x 0. 2 um. GC Column Format: Intuvo. Inner Diameter (ID): 0.25 mm. Length: 60 m. Length (m): 60 m. Polarity: High Polarity 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Capillary Column Phase: CP-Sil 88. Capillary Tubing: Fused Silica. Film Thickness: 0.2 um. GC Column Dim: 60 m x 0. 2 um. GC Column Format: Intuvo. Inner Diameter (ID): 0.25 mm. Length: 60 m. Length (m): 60 m. Polarity: High Polarity 1 * 1 Pce HEWLCP7487-INT 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou