Specifications:- Capillary Column Phase: Select Mineral Oil. Capillary Tubing: Fused Silica. Film Thickness: 0.10 um. GC Column Dim: 15 m x 0.1 um. GC Column Format: Intuvo. Inner Diameter (ID): 0.32 mm. Length: 15 m. Length (m): 15 m. Polarity: Low 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Capillary Column Phase: Select Mineral Oil. Capillary Tubing: Fused Silica. Film Thickness: 0.10 um. GC Column Dim: 15 m x 0.1 um. GC Column Format: Intuvo. Inner Diameter (ID): 0.32 mm. Length: 15 m. Length (m): 15 m. Polarity: Low 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Capillary Column Phase: Select Mineral Oil. Capillary Tubing: Fused Silica. Film Thickness: 0.10 um. GC Column Dim: 15 m x 0.1 um. GC Column Format: Intuvo. Inner Diameter (ID): 0.32 mm. Length: 15 m. Length (m): 15 m. Polarity: Low 1 * 1 Pce HEWLCP7491-INT 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou