Specifications:- Capillary Column Phase: CP-Sil 5 CB. Capillary Tubing: Fused Silica. Film Thickness: 5 um. GC Column Dim: 50 m x 5 um. GC Column Format: Intuvo. Inner Diameter (ID): 0.32 mm. Length: 50 m. Length (m): 50 m. Polarity: Low Polarity. Wi 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Capillary Column Phase: CP-Sil 5 CB. Capillary Tubing: Fused Silica. Film Thickness: 5 um. GC Column Dim: 50 m x 5 um. GC Column Format: Intuvo. Inner Diameter (ID): 0.32 mm. Length: 50 m. Length (m): 50 m. Polarity: Low Polarity. Wi 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Capillary Column Phase: CP-Sil 5 CB. Capillary Tubing: Fused Silica. Film Thickness: 5 um. GC Column Dim: 50 m x 5 um. GC Column Format: Intuvo. Inner Diameter (ID): 0.32 mm. Length: 50 m. Length (m): 50 m. Polarity: Low Polarity. Wi 1 * 1 Pce HEWLCP7690-INT 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou