Specifications:- Capillary Column Phase: CP-Wax 52 CB. Capillary Tubing: Fused Silica. Film Thickness: 0.2 um. GC Column Format: 7 i nch. Inner Diameter (ID): 0.2 mm. Length: 50 m. Polarity: High Polarity. With Smart Key: No. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Capillary Column Phase: CP-Wax 52 CB. Capillary Tubing: Fused Silica. Film Thickness: 0.2 um. GC Column Format: 7 i nch. Inner Diameter (ID): 0.2 mm. Length: 50 m. Polarity: High Polarity. With Smart Key: No. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Capillary Column Phase: CP-Wax 52 CB. Capillary Tubing: Fused Silica. Film Thickness: 0.2 um. GC Column Format: 7 i nch. Inner Diameter (ID): 0.2 mm. Length: 50 m. Polarity: High Polarity. With Smart Key: No. 1 * 1 Pce HEWLCP7785I 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou