Specifications:- Capillary Column Phase: Lowox. Capillary Tubing: Fused Silica. Film Thickness: 10 um. GC Column Format: 7 inch. GC Column Type: PLOT. Inner Diameter (ID): 0.53 mm. Length: 10 m. With Smart Key: No. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Capillary Column Phase: Lowox. Capillary Tubing: Fused Silica. Film Thickness: 10 um. GC Column Format: 7 inch. GC Column Type: PLOT. Inner Diameter (ID): 0.53 mm. Length: 10 m. With Smart Key: No. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Capillary Column Phase: Lowox. Capillary Tubing: Fused Silica. Film Thickness: 10 um. GC Column Format: 7 inch. GC Column Type: PLOT. Inner Diameter (ID): 0.53 mm. Length: 10 m. With Smart Key: No. 1 * 1 Pce HEWLCP8586 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou