Specifications:- Capillary Column Phase: VF-200ms. Capillary Tubing: Fused Silica. Film Thickness: 0.1 um. GC Column Format: 7 inch. GC Column Type: GC/MS. Inner Diameter (ID): 0.25 mm. Length: 60 m. Polarity: Mid Polarity. Temperature Range: 0 C- 3 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Capillary Column Phase: VF-200ms. Capillary Tubing: Fused Silica. Film Thickness: 0.1 um. GC Column Format: 7 inch. GC Column Type: GC/MS. Inner Diameter (ID): 0.25 mm. Length: 60 m. Polarity: Mid Polarity. Temperature Range: 0 C- 3 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Capillary Column Phase: VF-200ms. Capillary Tubing: Fused Silica. Film Thickness: 0.1 um. GC Column Format: 7 inch. GC Column Type: GC/MS. Inner Diameter (ID): 0.25 mm. Length: 60 m. Polarity: Mid Polarity. Temperature Range: 0 C- 3 1 * 1 Pce HEWLCP8869 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou