Specifications:- Capillary Column Phase: VF-1ms. Capillary Tubing: Fused Silica. Film Thickness: 0.25 um. GC Column Dim: 30 m x 0.25 um. GC Column Format: Intuvo. GC Column Type: GC/MS. Inner Diameter (ID): 0.25 mm. Length: 30 m. Length (m): 30 m. P 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Capillary Column Phase: VF-1ms. Capillary Tubing: Fused Silica. Film Thickness: 0.25 um. GC Column Dim: 30 m x 0.25 um. GC Column Format: Intuvo. GC Column Type: GC/MS. Inner Diameter (ID): 0.25 mm. Length: 30 m. Length (m): 30 m. P 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Capillary Column Phase: VF-1ms. Capillary Tubing: Fused Silica. Film Thickness: 0.25 um. GC Column Dim: 30 m x 0.25 um. GC Column Format: Intuvo. GC Column Type: GC/MS. Inner Diameter (ID): 0.25 mm. Length: 30 m. Length (m): 30 m. P 1 * 1 Pce HEWLCP8912-INT 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou