GC columns, J&W VF-17ms
Fournisseur: Agilent
CP9007
HEWLCP9007EA 1310 EUR InStock
HEWLCP9007
GC columns, J&W VF-17ms
Colonnes pour chromatographie
VF-17ms is a 50% phenyl/50% dimethylpolysiloxane, medium-polarity, low bleed column ideal for environmental and clinical research methods.
  • Proprietary deactivation technology and manufacturing process improves column stability, resulting in enhanced column-to-column repeatability and column lifetimes
  • Ideal for environmental and clinical methods
  • Ultralow bleed specification at 2 pA at 325 °C (0,25 mm × 30 m, 0,25 m)
J&W VF-17ms is a 50% phenyl/50% dimethylpolysiloxane, medium-polarity, low bleed column for increased sensitivity, accuracy and instrument uptime. VF-17ms is often referenced in environmental and clinical research methods. The use of deactivation technology improves column stability, resulting in improved repeatability and column lifetimes.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité