Specifications:- LC Application Compatibility: Bio-compatible. LC Module Type Compatibility: Detector. LC Supplies Brand: InfinityLa b. LC System Compatibility: 1290 infinity II Bio LC System. LC Use With Instrument: G7114A G7114B. Path Length: 10 mm 1 * 1 Pce
Fournisseur: Agilent
Specifications:- LC Application Compatibility: Bio-compatible. LC Module Type Compatibility: Detector. LC Supplies Brand: InfinityLa b. LC System Compatibility: 1290 infinity II Bio LC System. LC Use With Instrument: G7114A G7114B. Path Length: 10 mm 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- LC Application Compatibility: Bio-compatible. LC Module Type Compatibility: Detector. LC Supplies Brand: InfinityLa b. LC System Compatibility: 1290 infinity II Bio LC System. LC Use With Instrument: G7114A G7114B. Path Length: 10 mm 1 * 1 Pce HEWLG1314-60188 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou