Specifications:- Compatible Column ID: 0.1-0.25 mm. Compatible Column OuterDiameter (OD): 0.36-0.39 mm. Ferrule Material: Stainless steel. Ferrule Type: Capillary. GC Component Type: Ferrule. Inner Diameter (ID): 0.4 mm. Manufacturer Compatibility: A 1 * 10 Pce
Fournisseur: Agilent
Specifications:- Compatible Column ID: 0.1-0.25 mm. Compatible Column OuterDiameter (OD): 0.36-0.39 mm. Ferrule Material: Stainless steel. Ferrule Type: Capillary. GC Component Type: Ferrule. Inner Diameter (ID): 0.4 mm. Manufacturer Compatibility: A 1 * 10 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Compatible Column ID: 0.1-0.25 mm. Compatible Column OuterDiameter (OD): 0.36-0.39 mm. Ferrule Material: Stainless steel. Ferrule Type: Capillary. GC Component Type: Ferrule. Inner Diameter (ID): 0.4 mm. Manufacturer Compatibility: A 1 * 10 Pce HEWLG2855-28501 1 * 10 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou