Specifications:- Compatible Column ID: 0.53 mm. Compatible Column OuterDiameter (OD): 0.7 mm. Ferrule Material: Stainless steel. Fer rule Type: Capillary. GC Component Type: Ferrule. Inner Diameter (ID): 0.8 mm. Manufacturer Compatibility: Agilent. M 1 * 10 Pce
Fournisseur: Agilent
Specifications:- Compatible Column ID: 0.53 mm. Compatible Column OuterDiameter (OD): 0.7 mm. Ferrule Material: Stainless steel. Fer rule Type: Capillary. GC Component Type: Ferrule. Inner Diameter (ID): 0.8 mm. Manufacturer Compatibility: Agilent. M 1 * 10 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Compatible Column ID: 0.53 mm. Compatible Column OuterDiameter (OD): 0.7 mm. Ferrule Material: Stainless steel. Fer rule Type: Capillary. GC Component Type: Ferrule. Inner Diameter (ID): 0.8 mm. Manufacturer Compatibility: Agilent. M 1 * 10 Pce HEWLG2855-28503 1 * 10 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou