Specifications:- Fitting Usage: Nebulizer. Notes: Suits MicroMist nebulizer with PEEK ratchet nebulizer gas connector used with 8900 ICP-MS. Spectroscopy Instrument Compatibility: 8900. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Fitting Usage: Nebulizer. Notes: Suits MicroMist nebulizer with PEEK ratchet nebulizer gas connector used with 8900 ICP-MS. Spectroscopy Instrument Compatibility: 8900. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Fitting Usage: Nebulizer. Notes: Suits MicroMist nebulizer with PEEK ratchet nebulizer gas connector used with 8900 ICP-MS. Spectroscopy Instrument Compatibility: 8900. 1 * 1 Pce HEWLG3266-80017 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou