Specifications:- Chemical Compatibility: Aqueous Corrosive acids. Cone Care Package: No. Extraction Lens Type: s-lens. Interface Con e Base Material: Nickel-plated Copper. Interface Cone Tip Diameter: 12 mm. Interface Cone Tip Material: Platinum. Int 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Chemical Compatibility: Aqueous Corrosive acids. Cone Care Package: No. Extraction Lens Type: s-lens. Interface Con e Base Material: Nickel-plated Copper. Interface Cone Tip Diameter: 12 mm. Interface Cone Tip Material: Platinum. Int 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Chemical Compatibility: Aqueous Corrosive acids. Cone Care Package: No. Extraction Lens Type: s-lens. Interface Con e Base Material: Nickel-plated Copper. Interface Cone Tip Diameter: 12 mm. Interface Cone Tip Material: Platinum. Int 1 * 1 Pce HEWLG3280-67142 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou