Specifications:- Inner Diameter (ID): 2.1 mm. Packed Column Configuration: LVO. Packed Column Length: 9 ft (2.74 m). Packed Column M aterial: UltiMetal. Packed Column Mesh: 45/60. Packed Column Outer Diameter (OD): 0.125 in. Packed Column Support: Mo 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Inner Diameter (ID): 2.1 mm. Packed Column Configuration: LVO. Packed Column Length: 9 ft (2.74 m). Packed Column M aterial: UltiMetal. Packed Column Mesh: 45/60. Packed Column Outer Diameter (OD): 0.125 in. Packed Column Support: Mo 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Inner Diameter (ID): 2.1 mm. Packed Column Configuration: LVO. Packed Column Length: 9 ft (2.74 m). Packed Column M aterial: UltiMetal. Packed Column Mesh: 45/60. Packed Column Outer Diameter (OD): 0.125 in. Packed Column Support: Mo 1 * 1 Pce HEWLG3591-81224 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou