Specifications:- Inner Diameter (ID): 2 mm. Packed Column Configuration: LVO. Packed Column Length: 15 ft (4.57 m). Packed Column Ma terial: UltiMetal. Packed Column Mesh: 80/100. Packed Column Outer Diameter (OD): 0.125 in. Packed Column Phase: TCEP 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Inner Diameter (ID): 2 mm. Packed Column Configuration: LVO. Packed Column Length: 15 ft (4.57 m). Packed Column Ma terial: UltiMetal. Packed Column Mesh: 80/100. Packed Column Outer Diameter (OD): 0.125 in. Packed Column Phase: TCEP 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Inner Diameter (ID): 2 mm. Packed Column Configuration: LVO. Packed Column Length: 15 ft (4.57 m). Packed Column Ma terial: UltiMetal. Packed Column Mesh: 80/100. Packed Column Outer Diameter (OD): 0.125 in. Packed Column Phase: TCEP 1 * 1 Pce HEWLG3591-81229 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou