Specifications:- Inner Diameter (ID): 1 mm. Length: 4 m. Packed Column Material: UltiMetal. Packed Column Mesh: 80/100. Packed Colum n Outer Diameter (OD): 1/16 in. Packed Column Phase: Molsieve 13X. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Inner Diameter (ID): 1 mm. Length: 4 m. Packed Column Material: UltiMetal. Packed Column Mesh: 80/100. Packed Colum n Outer Diameter (OD): 1/16 in. Packed Column Phase: Molsieve 13X. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Inner Diameter (ID): 1 mm. Length: 4 m. Packed Column Material: UltiMetal. Packed Column Mesh: 80/100. Packed Colum n Outer Diameter (OD): 1/16 in. Packed Column Phase: Molsieve 13X. 1 * 1 Pce HEWLG3591-81231 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou