Specifications:- Chemical Compatibility: Aqueous Corrosive acids. Extraction Lens Type: x-lens. Interface Cone Type: Base. Skimmer B ase Material: Stainless steel. Spectroscopy Instrument Compatibility: 8900. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Chemical Compatibility: Aqueous Corrosive acids. Extraction Lens Type: x-lens. Interface Cone Type: Base. Skimmer B ase Material: Stainless steel. Spectroscopy Instrument Compatibility: 8900. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Chemical Compatibility: Aqueous Corrosive acids. Extraction Lens Type: x-lens. Interface Cone Type: Base. Skimmer B ase Material: Stainless steel. Spectroscopy Instrument Compatibility: 8900. 1 * 1 Pce HEWLG3666-60301 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou