Specifications:- Chemical Compatibility: Aqueous Corrosive acids Strong acids Organic solvents. Extraction Lens Type: s-lens. Interf ace Cone Type: Base. Skimmer Base Material: Brass. Spectroscopy Instrument Compatibility: 8900. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Chemical Compatibility: Aqueous Corrosive acids Strong acids Organic solvents. Extraction Lens Type: s-lens. Interf ace Cone Type: Base. Skimmer Base Material: Brass. Spectroscopy Instrument Compatibility: 8900. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Chemical Compatibility: Aqueous Corrosive acids Strong acids Organic solvents. Extraction Lens Type: s-lens. Interf ace Cone Type: Base. Skimmer Base Material: Brass. Spectroscopy Instrument Compatibility: 8900. 1 * 1 Pce HEWLG3666-60401 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou