Specifications:- Chemical Compatibility: Aqueous Corrosive acids Strong acids. Cone Care Package: No. Interface Cone Base Material: Nickel. Interface Cone Tip Material: Platinum. Interface Cone Type: Skimmer. Spectroscopy Instrument Compatibility: 89 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Chemical Compatibility: Aqueous Corrosive acids Strong acids. Cone Care Package: No. Interface Cone Base Material: Nickel. Interface Cone Tip Material: Platinum. Interface Cone Type: Skimmer. Spectroscopy Instrument Compatibility: 89 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Chemical Compatibility: Aqueous Corrosive acids Strong acids. Cone Care Package: No. Interface Cone Base Material: Nickel. Interface Cone Tip Material: Platinum. Interface Cone Type: Skimmer. Spectroscopy Instrument Compatibility: 89 1 * 1 Pce HEWLG3666-67411 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou