Specifications:- Capillary Column Phase: DB-1ht. Capillary Tubing: High Temperature Fused Silica. Film Thickness: 0.1 um. GC Column Format: 7 inch. Inner Diameter (ID): 0.25 mm. Length: 2 m. Length (m): 2 m. Polarity: Low Polarity. With Smart Key: No 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Capillary Column Phase: DB-1ht. Capillary Tubing: High Temperature Fused Silica. Film Thickness: 0.1 um. GC Column Format: 7 inch. Inner Diameter (ID): 0.25 mm. Length: 2 m. Length (m): 2 m. Polarity: Low Polarity. With Smart Key: No 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Capillary Column Phase: DB-1ht. Capillary Tubing: High Temperature Fused Silica. Film Thickness: 0.1 um. GC Column Format: 7 inch. Inner Diameter (ID): 0.25 mm. Length: 2 m. Length (m): 2 m. Polarity: Low Polarity. With Smart Key: No 1 * 1 Pce HEWLG3903-61006 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou