ISIS supplies for 7700
Fournisseur: Agilent
G4911-01201
HEWLG4911-01201EA 164 EUR InStock
HEWLG4911-01201
ISIS supplies for 7700
Matériel de chromatographie Chromatographie, Pièces de rechange
A range of high quality supplies and components for the Integrated Sample Introduction System (ISIS) for use with the 7700 ICP-MS range.
  • A variety of joints are available in tee, cross or union form
Agilent's ISIS enhances the functionality and flexibility of the Agilent 7700 series ICP-MS. ISIS features high precision peristaltic pumps and an optional switching valve to allow complete flexibility of configuration. The ISIS hardware is built into the 7700 chassis and close-coupled to increase productivity through high-speed sample uptake and rapid washout. To enhance the capabilities that the system offers and maintain performance even after heavy use, Agilent provides a range of replacement components and other supplies.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité