Specifications:- LC Application Compatibility: Bio-inert. LC Module Type Compatibility: Autosampler. LC System Compatibility: 1260 I nfinity II Bio-Inert LC System. LC Use With Instrument: G5668A. Needle Material: Ceramic. Technique: LC. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- LC Application Compatibility: Bio-inert. LC Module Type Compatibility: Autosampler. LC System Compatibility: 1260 I nfinity II Bio-Inert LC System. LC Use With Instrument: G5668A. Needle Material: Ceramic. Technique: LC. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- LC Application Compatibility: Bio-inert. LC Module Type Compatibility: Autosampler. LC System Compatibility: 1260 I nfinity II Bio-Inert LC System. LC Use With Instrument: G5668A. Needle Material: Ceramic. Technique: LC. 1 * 1 Pce HEWLG5668-87200 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou