Alignment interfaces and cassettes
Fournisseur: Agilent
G7100-60400
HEWLG7100-60400EA 718 EUR InStock
HEWLG7100-60400
Alignment interfaces and cassettes
Matériel de chromatographie Chromatographie, Pièces de rechange
These alignment interfaces are an integral part of the Agilent diode-array detection system.
  • The quick-change cassette allows capillary exchange in less than a minute
  • The cassette and interfaces accept all commercially available capillaries (~365 µm od)
These interfaces contain optical slits which are precisely matched to the capillary inner diameter for optimized sensitivity and linear detection range. In combination with the capillary cassette, alignment interfaces simplify capillary exchange, protect the fragile detection window and ensure exact alignment of the window in the detector.
Attention: The color code of the alignment interface must match with the color code of the capillary's built-in alignment stopper.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité