Specifications:- Heat Exchanger Material: PEEK/SST. LC Application Compatibility: Bio-inert. LC Module Type Compatibility: Column Co mpartment. LC Supplies Brand: InfinityLab. LC System Compatibility: 1260 Infinity II Bio-Inert LC System. LC Use With 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Heat Exchanger Material: PEEK/SST. LC Application Compatibility: Bio-inert. LC Module Type Compatibility: Column Co mpartment. LC Supplies Brand: InfinityLab. LC System Compatibility: 1260 Infinity II Bio-Inert LC System. LC Use With 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Heat Exchanger Material: PEEK/SST. LC Application Compatibility: Bio-inert. LC Module Type Compatibility: Column Co mpartment. LC Supplies Brand: InfinityLab. LC System Compatibility: 1260 Infinity II Bio-Inert LC System. LC Use With 1 * 1 Pce HEWLG7116-60009 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou