Specifications:- Heat Exchanger Material: MP35N. LC Application Compatibility: Bio-compatible. LC Module Type Compatibility: Column Compartment. LC System Compatibility: 1290 infinity II Bio LC System. LC Use With Instrument: G7116B G7116A. Volume: 1 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Heat Exchanger Material: MP35N. LC Application Compatibility: Bio-compatible. LC Module Type Compatibility: Column Compartment. LC System Compatibility: 1290 infinity II Bio LC System. LC Use With Instrument: G7116B G7116A. Volume: 1 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Heat Exchanger Material: MP35N. LC Application Compatibility: Bio-compatible. LC Module Type Compatibility: Column Compartment. LC System Compatibility: 1290 infinity II Bio LC System. LC Use With Instrument: G7116B G7116A. Volume: 1 1 * 1 Pce HEWLG7116-60071 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou