Specifications:- LC Application Compatibility: Analytical. LC Module Type Compatibility: Pump. LC System Compatibility: 1290 Infinit y LC System 1290 Infinity II LC System. LC Use With Instrument: G4220A G4220B G7120A. Mixer Material: Stainless Steel 1 * 1 Pce
Fournisseur: Agilent
Specifications:- LC Application Compatibility: Analytical. LC Module Type Compatibility: Pump. LC System Compatibility: 1290 Infinit y LC System 1290 Infinity II LC System. LC Use With Instrument: G4220A G4220B G7120A. Mixer Material: Stainless Steel 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- LC Application Compatibility: Analytical. LC Module Type Compatibility: Pump. LC System Compatibility: 1290 Infinit y LC System 1290 Infinity II LC System. LC Use With Instrument: G4220A G4220B G7120A. Mixer Material: Stainless Steel 1 * 1 Pce HEWLG7120-68380 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou